COM448633572 Combi-Blocks, Inc.

6-(Tetramethyl-1,3,2-dioxaborolan-2-yl)-2,3-dihydro-1-benzopyran-4-one

  • MDL number MFCD27936765
  • Cas Number : 1421659-47-3
  • Molecular Weight : 274.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-2894

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-2894
Availability details for
Quantity
  • Availability
  • Add to Cart