COM448633573 Combi-Blocks, Inc.

5-(Tetramethyl-1,3,2-dioxaborolan-2-yl)-[1,2,4]triazolo[1,5-a]pyridin-2-amine

  • MDL number MFCD27936766
  • Molecular Weight : 260.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-2895

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-2895
Availability details for
Quantity
  • Availability
  • Add to Cart