COM448633585 Combi-Blocks, Inc.

7-(Tetramethyl-1,3,2-dioxaborolan-2-yl)-3,4-dihydro-2H-naphthalen-1-one

  • MDL number MFCD27936771
  • Cas Number : 1135871-91-8
  • Molecular Weight : 272.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-2945

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-2945
Availability details for
Quantity
  • Availability
  • Add to Cart