COM448633586 Combi-Blocks, Inc.

6-Chloro-3-(tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazin-2-amine

  • MDL number MFCD27936772
  • Molecular Weight : 255.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-2946

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-2946
Availability details for
Quantity
  • Availability
  • Add to Cart