COM448633588 Combi-Blocks, Inc.

4-Amino-2-(tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile

  • MDL number MFCD24040038
  • Cas Number : 1421322-57-7
  • Molecular Weight : 244.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-2949

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-2949
Availability details for
Quantity
  • Availability
  • Add to Cart