COM448633591 Combi-Blocks, Inc.

1-[2-(Tetramethyl-1,3,2-dioxaborolan-2-yl)ethyl]pyrrolidine-2,5-dione

  • MDL number MFCD27936775
  • Molecular Weight : 253.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-2961

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-2961
Availability details for
Quantity
  • Availability
  • Add to Cart