COM448633595 Combi-Blocks, Inc.

1-(4'-Fluorophenyl)ethanone-2-boronic acid pinacol ester

  • MDL number MFCD25541627
  • Molecular Weight : 264.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-2965

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-2965
Availability details for
Quantity
  • Availability
  • Add to Cart