COM448633598 Combi-Blocks, Inc.

3-Nitro-4-(propylamino)-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile

  • MDL number MFCD27936780
  • Molecular Weight : 331.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-2968

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-2968
Availability details for
Quantity
  • Availability
  • Add to Cart