COM448633599 Combi-Blocks, Inc.

2-[2-Methanesulfonyl-4-nitro-5-(trifluoromethoxy)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

  • MDL number MFCD27936781
  • Molecular Weight : 411.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-2969

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-2969
Availability details for
Quantity
  • Availability
  • Add to Cart