COM448633606 Combi-Blocks, Inc.

4-(Methylamino)-3-nitro-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile

  • MDL number MFCD27936788
  • Molecular Weight : 303.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-2976

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-2976
Availability details for
Quantity
  • Availability
  • Add to Cart