COM448633610 Combi-Blocks, Inc.

Methyl 7-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-1,2,3-benzotriazole-5-carboxylate

  • MDL number MFCD27936792
  • Molecular Weight : 303.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-2980

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-2980
Availability details for
Quantity
  • Availability
  • Add to Cart