COM448633612 Combi-Blocks, Inc.

Methyl 1-methyl-7-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3-benzodiazole-5-carboxylate

  • MDL number MFCD27936794
  • Molecular Weight : 316.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-2982

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-2982
Availability details for
Quantity
  • Availability
  • Add to Cart