COM448633616 Combi-Blocks, Inc.

4-(Ethylamino)-3-nitro-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile

  • MDL number MFCD27936798
  • Molecular Weight : 317.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-2986

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-2986
Availability details for
Quantity
  • Availability
  • Add to Cart