COM448633642 Combi-Blocks, Inc.

Ethyl 3-amino-4-(methylamino)-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

  • MDL number MFCD27936812
  • Molecular Weight : 320.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-3022

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-3022
Availability details for
Quantity
  • Availability
  • Add to Cart