COM448633653 Combi-Blocks, Inc.

3-(N-Ethylaminocarbonyl)phenylboronic acid pinacol ester

  • MDL number MFCD22493890
  • Cas Number : 943911-67-9
  • Molecular Weight : 275.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-3057

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-3057
Availability details for
Quantity
  • Availability
  • Add to Cart