COM448633664 Combi-Blocks, Inc.

3-Amino-4-(propylamino)-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile

  • MDL number MFCD27936816
  • Molecular Weight : 301.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-3133

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-3133
Availability details for
Quantity
  • Availability
  • Add to Cart