COM448633665 Combi-Blocks, Inc.

7-Methyl-6-(tetramethyl-1,3,2-dioxaborolan-2-yl)-3H-[1,2,3]triazolo[4,5-b]pyridine

  • MDL number MFCD27936817
  • Molecular Weight : 260.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-3134

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-3134
Availability details for
Quantity
  • Availability
  • Add to Cart