COM448633677 Combi-Blocks, Inc.

1-Propyl-7-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1,2,3-benzotriazole-5-carboxylic acid

  • MDL number MFCD27936828
  • Molecular Weight : 331.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-3146

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-3146
Availability details for
Quantity
  • Availability
  • Add to Cart