COM448633680 Combi-Blocks, Inc.

Ethyl 1-propyl-7-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1,2,3-benzotriazole-5-carboxylate

  • MDL number MFCD27936831
  • Molecular Weight : 359.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-3149

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-3149
Availability details for
Quantity
  • Availability
  • Add to Cart