COM448633685 Combi-Blocks, Inc.

7-Fluoro-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1,4-dihydroquinoxaline-2,3-dione

  • MDL number MFCD27936836
  • Molecular Weight : 306.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-3184

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-3184
Availability details for
Quantity
  • Availability
  • Add to Cart