COM448633735 Combi-Blocks, Inc.

2-Chloro-5-methoxycarbonylphenylboronic acid pinacol ester

  • MDL number MFCD16996247
  • Cas Number : 710350-72-4
  • Molecular Weight : 296.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-3528

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-3528
Availability details for
Quantity
  • Availability
  • Add to Cart