COM448633883 Combi-Blocks, Inc.

3-Fluoro-2,4-dimethoxyphenylboronic acid pinacol ester

  • MDL number MFCD28122693
  • Cas Number : 2377609-46-4
  • Molecular Weight : 282.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-4286

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-4286
Availability details for
Quantity
  • Availability
  • Add to Cart