COM448633936 Combi-Blocks, Inc.

5-BOC-Amino-2-chlorophenylboronic acid pinacol ester

  • MDL number MFCD23379564
  • Cas Number : 1080573-28-9
  • Molecular Weight : 353.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-4573

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-4573
Availability details for
Quantity
  • Availability
  • Add to Cart