COM448633978 Combi-Blocks, Inc.

3-[(N-BOC-Azetidin-3-yl)sulfanyl]phenylboronic acid, pinacol ester

  • MDL number MFCD27936842
  • Cas Number : 2377607-25-3
  • Molecular Weight : 391.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-4797

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-4797
Availability details for
Quantity
  • Availability
  • Add to Cart