COM448633979 Combi-Blocks, Inc.

4-Fluoro-3-(pyrrolidinomethyl)phenylboronic acid pinacol ester

  • MDL number MFCD27936843
  • Cas Number : 2377607-32-2
  • Molecular Weight : 305.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-4798

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-4798
Availability details for
Quantity
  • Availability
  • Add to Cart