COM448633981 Combi-Blocks, Inc.

3-(N-Diethylamino)methyl-4-fluorophenylboronic acid pinacol ester

  • MDL number MFCD27981319
  • Cas Number : 2377609-50-0
  • Molecular Weight : 307.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-4800

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-4800
Availability details for
Quantity
  • Availability
  • Add to Cart