COM448633986 Combi-Blocks, Inc.

1-[4-(Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]pent-4-en-1-one

  • MDL number MFCD27936847
  • Cas Number : 2377606-47-6
  • Molecular Weight : 286.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-4809

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-4809
Availability details for
Quantity
  • Availability
  • Add to Cart