COM448633987 Combi-Blocks, Inc.

3-Cyano-4-fluoro-5-methoxyphenylboronic acid pinacol ester

  • Cas Number : 1383985-45-2
  • Molecular Weight : 277.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-4814

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-4814
Availability details for
Quantity
  • Availability
  • Add to Cart