COM448633991 Combi-Blocks, Inc.

4-(N-BOC-Amino)-5-(methoxycarbonyl)thiophene-2-boronic acid, pinacol ester

  • MDL number MFCD28014041
  • Molecular Weight : 383.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 98
Storage temp. Freezer

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-4841
Availability details for
Quantity
  • Availability
  • Add to Cart