COM448633992 Combi-Blocks, Inc.

1-Methyl-4-(methoxycarbonyl)pyrrole-2-boronic acid pinacol ester

  • MDL number MFCD28096644
  • Molecular Weight : 265.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Freezer
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-4846

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-4846
Availability details for
Quantity
  • Availability
  • Add to Cart