COM448633997 Combi-Blocks, Inc.

3,4-Dimethyl-5-propoxyphenylboronic acid pinacol ester

  • MDL number MFCD28096646
  • Molecular Weight : 290.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-4879

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-4879
Availability details for
Quantity
  • Availability
  • Add to Cart