COM448633998 Combi-Blocks, Inc.

3,4-Dimethoxy-5-(3-methoxypropoxy)phenylboronic acid pinacol ester

  • Molecular Weight : 320.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-4880

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-4880
Availability details for
Quantity
  • Availability
  • Add to Cart