COM448634000 Combi-Blocks, Inc.

3,4-Dichloro-5-propoxyphenylboronic acid pinacol ester

  • MDL number MFCD28096649
  • Molecular Weight : 331.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-4882

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-4882
Availability details for
Quantity
  • Availability
  • Add to Cart