COM448634001 Combi-Blocks, Inc.

2-(3,4-Dichloro-5-isopropoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

  • MDL number MFCD28096650
  • Molecular Weight : 331.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-4883

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-4883
Availability details for
Quantity
  • Availability
  • Add to Cart