COM448634002 Combi-Blocks, Inc.

2-(3-Ethoxy-4,5-dimethylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

  • MDL number MFCD28096651
  • Cas Number : 2377609-57-7
  • Molecular Weight : 276.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-4884

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-4884
Availability details for
Quantity
  • Availability
  • Add to Cart