COM448634003 Combi-Blocks, Inc.

1-BOC-3-Cyclopropyl-1H-pyrazole-4-boronic acid, pinacol ester

  • MDL number MFCD28122694
  • Molecular Weight : 334.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-4890

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-4890
Availability details for
Quantity
  • Availability
  • Add to Cart