COM448634005 Combi-Blocks, Inc.

1-BOC-4-(Methoxycarbonyl)indole-6-boronic acid, pinacol ester

  • MDL number MFCD28101648
  • Molecular Weight : 401.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Freezer
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-4898

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-4898
Availability details for
Quantity
  • Availability
  • Add to Cart