COM448634007 Combi-Blocks, Inc.

2-Fluoro-3-(methylcarbamoyl)phenylboronic acid pinacol ester

  • MDL number MFCD18730367
  • Molecular Weight : 279.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-4905

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-4905
Availability details for
Quantity
  • Availability
  • Add to Cart