COM448634017 Combi-Blocks, Inc.

3-Methoxy-5-(methoxycarbonylmethyl)phenylboronic acid pinacol ester

  • MDL number MFCD28143943
  • Molecular Weight : 306.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-4986

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-4986
Availability details for
Quantity
  • Availability
  • Add to Cart