COM448634020 Combi-Blocks, Inc.

1-[4-(Tetramethyl-1,3,2-dioxaborolan-2-yl)piperidin-1-yl]propan-1-one

  • MDL number MFCD28143946
  • Molecular Weight : 267.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-4990

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-4990
Availability details for
Quantity
  • Availability
  • Add to Cart