COM448634027 Combi-Blocks, Inc.

4-(Tetramethyl-1,3,2-dioxaborolan-2-yl)-1-[(thiophen-2-yl)carbonyl]piperidine

  • MDL number MFCD28143953
  • Cas Number : 2016819-12-6
  • Molecular Weight : 321.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-4997

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-4997
Availability details for
Quantity
  • Availability
  • Add to Cart