COM448634029 Combi-Blocks, Inc.

N-Tosylpiperidine-4-boronic acid pinacol ester

  • MDL number MFCD28166403
  • Cas Number : 1382358-37-3
  • Molecular Weight : 365.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Freezer
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-4999

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-4999
Availability details for
Quantity
  • Availability
  • Add to Cart