COM448634077 Combi-Blocks, Inc.

3-Chloro-4-cyanophenylboronic acid pinacol ester

  • MDL number MFCD16996249
  • Cas Number : 548797-51-9
  • Molecular Weight : 263.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-5248

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-5248
Availability details for
Quantity
  • Availability
  • Add to Cart