COM448634138 Combi-Blocks, Inc.

3-Bromo-2-methylpyridine-5-boronic acid pinacol ester

  • MDL number MFCD16995186
  • Cas Number : 2223051-53-2
  • Molecular Weight : 298.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-5508

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-5508
Availability details for
Quantity
  • Availability
  • Add to Cart