COM448634232 Combi-Blocks, Inc.

2-(2-Methoxy-5-nitrophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

  • MDL number MFCD20327978
  • Cas Number : 677746-34-8
  • Molecular Weight : 279.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-5765

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-5765
Availability details for
Quantity
  • Availability
  • Add to Cart