COM448634252 Combi-Blocks, Inc.

2-(N-BOC-Aminomethyl)-5-nitrophenylboronic acid pinacol ester

  • MDL number MFCD28143955
  • Molecular Weight : 378.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-5849

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-5849
Availability details for
Quantity
  • Availability
  • Add to Cart