COM448634261 Combi-Blocks, Inc.

3-chloro-2-(2-methylpropoxy)-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

  • MDL number MFCD24368939
  • Cas Number : 1387634-81-2
  • Molecular Weight : 311.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-5878

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-5878
Availability details for
Quantity
  • Availability
  • Add to Cart