COM448634271 Combi-Blocks, Inc.

2-Methylaminopyrimidine-5-boronic acid, pinacol ester

  • MDL number MFCD11878345
  • Cas Number : 904326-88-1
  • Molecular Weight : 235.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-5920

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-5920
Availability details for
Quantity
  • Availability
  • Add to Cart