COM448634330 Combi-Blocks, Inc.

2-bromo-3-fluoro-4-(tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

  • MDL number MFCD18915056
  • Molecular Weight : 301.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-5997

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?PN-5997
Availability details for
Quantity
  • Availability
  • Add to Cart