COM448634508 Combi-Blocks, Inc.

2-Chloro-4-pyrrolidinomethylphenylboronic acid, pinacol ester

  • MDL number MFCD18087735
  • Cas Number : 1256360-54-9
  • Molecular Weight : 321.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?PN-6670

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?PN-6670
Availability details for
Quantity
  • Availability
  • Add to Cart